siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175236.4 | NP_780445.1 | 5 | 256 | ||
GenBank mRNA | AK018241.1 | - | 5 | 230 | ||
AK038853.1 | - | 5 | 223 | |||
AK046780.1 | - | 5 | 182 | |||
AK050178.1 | - | 4 | 180 | |||
AK084104.1 | - | 183 | ||||
BC026584.1 | - | 5 | 214 | 4972323 | ||
CA467848.1 | - | 5 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
