siRNA Details
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133763.1 | NP_598524.1 | 2 | 160 | ||
GenBank mRNA | AB045974.1 | - | 2 | 153 | ||
AK018299.1 | - | 2 | 187 | |||
AK145239.1 | - | 2 | 186 | |||
AK168315.1 | - | 2 | 162 | |||
AK168357.1 | - | 2 | 180 | |||
BC003486.1 | - | 2 | 160 | 2811159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
