siRNA Details
Gene Symbol: Tax1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029564.2 | NP_083840.1 | 2 | 273 | ||
GenBank mRNA | AK004963.1 | - | 2 | 274 | ||
AK085679.1 | - | 106 | ||||
AK150816.1 | - | 2 | 106 | |||
AK168533.1 | - | 2 | 147 | |||
AK170485.1 | - | 2 | 139 | |||
BC008166.1 | - | 2 | 94 | 3257323 | ||
BC094314.1 | - | 2 | 79 | 5715687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
