siRNA Details
Gene Symbol: Trp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 1066 | ||
GenBank mRNA | AK011574.2 | - | 2 | 1027 | ||
AK028411.1 | - | 2 | 1067 | |||
AK046261.1 | - | 2 | 1044 | |||
AK049291.1 | - | 2 | 1051 | |||
AK160279.1 | - | 2 | 1046 | |||
BC017155.1 | - | 2 | 1084 | 4910610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
