siRNA Details
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172426.2 | NP_766014.1 | 4, 5 | 1462 | ||
GenBank mRNA | AK012771.1 | - | 268 | |||
AK083127.1 | - | 4, 5 | 1346 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031743, AF021923.
Gene Ontology Categories:
