siRNA Details
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110240.1 | NP_001103710.1 | 7 | 1725 | ||
NM_172426.2 | NP_766014.1 | 8 | 1815 | |||
GenBank mRNA | AK012771.1 | - | 621 | |||
AK083127.1 | - | 8 | 1699 | |||
BC115867.1 | - | 7 | 1727 | 40081382 | ||
BC116637.1 | - | 6 | 1541 | 40081381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
