siRNA Details
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029586.2 | NP_083862.1 | 3 | 370 | ||
NM_029606.3 | NP_083882.3 | 26 | 2917 | |||
NM_145688.2 | NP_663721.2 | 5 | 592 | |||
GenBank mRNA | AK005611.1 | - | 3 | 371 | ||
AK006504.1 | - | 5 | 578 | |||
BC005723.1 | - | 5 | 404 | |||
BC048677.1 | - | 5 | 564 | |||
BC086808.1 | - | 20 | 2157 | 30632964 | ||
CA464637.1 | - | 5 | 594 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
