siRNA Details
Gene Symbol: 1700012A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029587.2 | NP_083863.1 | 3 | 513 | ||
GenBank mRNA | AK005884.1 | - | 3 | 513 | ||
BC059718.1 | - | 3 | 212 | |||
BC087908.1 | - | 3 | 496 | 6774051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
