siRNA Details
Gene Symbol: Zfp735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126489.2 | NP_001119961.1 | 3, 4 | 425 | ||
GenBank mRNA | AK145370.1 | - | 3, 4 | 426 | ||
BC137974.1 | - | 3, 4 | 355 | 8860993 | ||
BY735607.1 | - | 3, 4 | 426 |
