siRNA Details
Gene Symbol: Il31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029594.1 | NP_083870.1 | 2 | 176 | ||
GenBank mRNA | AK005939.1 | - | 2 | 346 | ||
AY509149.1 | - | 2 | 176 | |||
BC117082.1 | - | 2 | 175 | 40126401 | ||
BC117084.1 | - | 2 | 175 | 40126403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
