siRNA Details
Gene Symbol: 1700019B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029598.1 | NP_083874.1 | 8 | 1353 | ||
GenBank mRNA | AK006106.1 | - | 5 | 598 | ||
BC050809.1 | - | 8 | 1348 | 6771839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
