siRNA Details
Gene Symbol: Sun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006225.1 | - | 8 | 554 | ||
AY307077.1 | - | 8 | 573 | |||
FJ667498.1 | - | 9 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
