siRNA Details
Gene Symbol: Ift43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199843.1 | NP_001186772.1 | 2 | 131 | ||
NM_029601.3 | NP_083877.1 | 2 | 131 | |||
GenBank mRNA | AK006115.1 | - | 2 | 129 | ||
BC034668.1 | - | 2 | 69 | |||
BC038915.1 | - | 2 | 120 | 5148723 | ||
CF751010.1 | - | 2 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
