siRNA Details
Gene Symbol: Pifo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200028.1 | NP_001186957.1 | 3 | 385 | ||
NM_029604.3 | NP_083880.2 | 3 | 337 | |||
GenBank mRNA | AK006407.1 | - | 3 | 338 | ||
AW519725.1 | - | 2 | 49 | |||
BC049757.1 | - | 3 | 386 | 6773491 | ||
HM237137.1 | - | 3 | 227 | |||
HM237138.1 | - | 3 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
