siRNA Details
Gene Symbol: Daam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008231.2 | NP_001008232.2 | 21 | 2816 | ||
GenBank mRNA | AK009382.1 | - | 6 | 638 | ||
AK122266.1 | - | 19 | 5734 | |||
AY426536.1 | - | 21 | 2845 | |||
BC037720.1 | - | 5 | 537 |
