siRNA Details
Gene Symbol: Daam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008231.2 | NP_001008232.2 | 26 | 3402 | ||
GenBank mRNA | AK009382.1 | - | 11 | 1224 | ||
AK122266.1 | - | 24 | 6320 | |||
AY426536.1 | - | 26 | 3431 | |||
BC037720.1 | - | 10 | 1123 |
