siRNA Details
Gene Symbol: Lmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029624.4 | NP_083900.3 | 10 | 1485 | ||
GenBank mRNA | AK010312.1 | - | 10 | 1456 | ||
AK041659.1 | - | 8 | 1152 | |||
AK154622.1 | - | 10 | 1482 | |||
AK169983.1 | - | 10 | 1483 | |||
BC020104.1 | - | 10 | 1492 | 3963699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
