siRNA Details
Gene Symbol: Paqr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023824.3 | NP_076313.1 | 1 | 362 | ||
GenBank mRNA | AB041564.1 | - | 1 | 283 | ||
AK005142.1 | - | 1 | 385 | |||
AK033625.1 | - | 1 | 380 | |||
AK044092.1 | - | 1 | 182 | |||
AK044486.1 | - | 1 | 385 | |||
AK152676.1 | - | 1 | 361 | |||
AY424293.1 | - | 1 | 313 | |||
BC011185.1 | - | 1 | 362 | 4196033 | ||
BC026921.1 | - | 1 | 313 | 4500871 |

More Information
Other Species Targeted

Gene Ontology Categories:
