siRNA Details
Gene Symbol: Lsm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133939.1 | NP_598700.1 | 3 | 209 | ||
GenBank mRNA | AK008067.1 | - | 3 | 191 | ||
AK088664.1 | - | 3 | 170 | |||
AK154446.1 | - | 1074 | ||||
AK167674.1 | - | 141 | ||||
BC019458.1 | - | 3 | 209 | 4024697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
