siRNA Details
Gene Symbol: Cln6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033175.2 | NP_001028347.1 | 7 | 930 | ||
GenBank mRNA | AK154414.1 | - | 7 | 931 | ||
AK154551.1 | - | 7 | 924 | |||
AK211299.1 | - | 3 | 370 | |||
AK216712.1 | - | 3 | 370 | |||
BC116794.1 | - | 6 | 756 | |||
BC116798.1 | - | 6 | 756 | |||
BC144722.1 | - | 6 | 815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
