siRNA Details
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029648.6 | NP_083924.2 | 2 | 564 | ||
GenBank mRNA | AK011404.2 | - | 2 | 543 | ||
AK075914.1 | - | 2 | 565 | |||
AK162079.1 | - | 2 | 516 | |||
BC048505.1 | - | 3 | 191 | |||
BC061459.1 | - | 363 | 6829614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
