siRNA Details
Gene Symbol: Tmem101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029649.2 | NP_083925.2 | 2 | 222 | ||
GenBank mRNA | AK012106.1 | - | 2 | 176 | ||
AK041154.1 | - | 2 | 181 | |||
AK049378.1 | - | 2 | 204 | |||
AK140260.1 | - | 2 | 223 | |||
BC011109.1 | - | 2 | 192 | 2654288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
