siRNA Details
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029669.4 | NP_083945.1 | 7 | 878 | ||
GenBank mRNA | AK012521.1 | - | 7 | 831 | ||
AK030483.1 | - | 7 | 856 | |||
BC026452.1 | - | 7 | 840 | 4460764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
