siRNA Details
Gene Symbol: 1700112E06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028275.1 | NP_082551.1 | 4 | 522 | ||
GenBank mRNA | AK007175.1 | - | 4 | 526 | ||
AK012226.1 | - | 2 | 347 | |||
AK153491.1 | - | 3 | 387 | |||
BY516672.1 | - | 67 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
