siRNA Details
Gene Symbol: 1700123K08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029693.2 | NP_083969.1 | 5 | 703 | ||
GenBank mRNA | AK007254.1 | - | 5 | 704 | ||
BC147525.1 | - | 5 | 691 | 9056517 | ||
BC147536.1 | - | 5 | 691 | 9056527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
