siRNA Details
Gene Symbol: Cfap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163638.1 | NP_001157110.1 | 3 | 297 | ||
NM_001163639.1 | NP_001157111.1 | 3 | 297 | |||
NM_029698.1 | NP_083974.1 | 3 | 235 | |||
GenBank mRNA | AK017232.1 | - | 3 | 298 | ||
BC138269.1 | - | 3 | 294 | 8861288 | ||
BC145225.1 | - | 3 | 294 | 9053777 | ||
BC145226.1 | - | 3 | 294 | 9053778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
