siRNA Details
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165991.1 | NP_001159463.1 | 5 | 505 | ||
NM_001165992.1 | NP_001159464.1 | 5 | 486 | |||
NM_001165995.1 | NP_001159467.1 | 4 | 392 | |||
NM_029702.4 | NP_083978.3 | 5 | 499 | |||
GenBank mRNA | AJ413953.1 | - | 5 | 452 | ||
AK005174.1 | - | 5 | 447 | |||
AK044759.1 | - | 5 | 457 | |||
AK050136.1 | - | 2 | 1357 | |||
AK083490.1 | - | 1 | 2882 | |||
AK180346.1 | - | 2 | 104 | |||
AK185500.1 | - | 2 | 105 | |||
BC010713.1 | - | 5 | 446 | 2649462 | ||
BC046782.1 | - | 5 | 437 | 5720540 | ||
BC046786.1 | - | 5 | 437 | 6416321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
