siRNA Details
Gene Symbol: Clasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293300.1 | NP_001280229.1 | 7, 8 | 1393 | ||
NM_001293301.1 | NP_001280230.1 | 7, 8 | 1393 | |||
NM_029709.2 | NP_083985.2 | 7, 8 | 1393 | |||
GenBank mRNA | AJ276962.1 | - | 7, 8 | 993 | ||
AK031181.1 | - | 7, 8 | 995 | |||
AK042402.1 | - | 7, 8 | 1397 | |||
AK080782.1 | - | 7, 8 | 1109 | |||
BC094432.1 | - | 6, 7 | 934 | 30674963 | ||
BC157938.1 | - | 7, 8 | 995 | |||
BC158064.1 | - | 7, 8 | 996 | 9007363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
