siRNA Details
Gene Symbol: 1700039E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033176.1 | NP_001028348.1 | 2 | 268 | ||
GenBank mRNA | AK133083.1 | - | 2 | 268 | ||
BC147591.1 | - | 2 | 365 | 9056581 | ||
BC147593.1 | - | 2 | 365 | 9056583 | ||
BC172112.1 | - | 2 | 365 |
