siRNA Details
Gene Symbol: Catsperg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029714.3 | NP_083990.2 | 2 | 159 | ||
GenBank mRNA | AK006911.1 | - | 2 | 150 | ||
AK077059.1 | - | 2 | 141 | |||
AK138347.1 | - | 2 | 159 | |||
BC144969.1 | - | 2 | 138 | |||
BY724013.1 | - | 2 | 159 | |||
GQ225581.1 | - | 1 | 60 |
Gene Symbol: Catsperg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164658.1 | NP_001158130.1 | 2 | 189 | ||
GenBank mRNA | AK039202.1 | - | 2 | 194 | ||
AK042364.1 | - | 2 | 121 | |||
BB633050.1 | - | 2 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
