siRNA Details
Gene Symbol: Dapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029723.3 | NP_083999.1 | 3, 4 | 268 | ||
GenBank mRNA | AK009567.1 | - | 3, 4 | 265 | ||
AK133382.1 | - | 384 | ||||
BC052745.1 | - | 403 | ||||
BC116916.1 | - | 3, 4 | 258 | 40126235 | ||
BC116920.1 | - | 3, 4 | 258 | 40126239 | ||
BY786476.1 | - | 3, 4 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
