siRNA Details
Gene Symbol: Trdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029726.2 | NP_084002.2 | 7 | 829 | ||
GenBank mRNA | AF223415.1 | - | 7 | 772 | ||
AF223416.1 | - | 7 | 772 | |||
AF223417.1 | - | 7 | 751 | |||
BC034343.1 | - | 7 | 752 | 3589077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
