siRNA Details
Gene Symbol: 2010005H15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029733.3 | NP_084009.1 | 5 | 413 | ||
GenBank mRNA | AK008122.1 | - | 5 | 508 | ||
BC125396.1 | - | 5 | 413 | 40129911 | ||
BC125422.1 | - | 5 | 413 | 40129937 | ||
BC145264.1 | - | 4 | 351 | 9053817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
