siRNA Details
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282118.1 | NP_001269047.1 | 2 | 217 | ||
NM_001282119.1 | NP_001269048.1 | 2 | 139 | |||
NM_001282120.1 | NP_001269049.1 | 2 | 495 | |||
NM_133767.3 | NP_598528.2 | 2 | 495 | |||
GenBank mRNA | AK010766.1 | - | 2 | 440 | ||
AK136344.1 | - | 2 | 179 | |||
AK146961.1 | - | 2 | 435 | |||
AK160724.1 | - | 2 | 451 | |||
BC016590.1 | - | 2 | 71 | 3585359 | ||
BC066819.1 | - | 2 | 389 | |||
CK620846.1 | - | 2 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
