siRNA Details
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282118.1 | NP_001269047.1 | 11 | 1795 | ||
NM_001282119.1 | NP_001269048.1 | 11 | 1717 | |||
NM_001282120.1 | NP_001269049.1 | 11 | 2159 | |||
NM_133767.3 | NP_598528.2 | 11 | 2073 | |||
GenBank mRNA | AK054313.1 | - | 2 | 947 | ||
AK136344.1 | - | 11 | 1757 | |||
AK146961.1 | - | 11 | 2013 | |||
BC016590.1 | - | 11 | 1649 | 3585359 | ||
BC066819.1 | - | 11 | 2053 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
