siRNA Details
Gene Symbol: Mzt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175245.4 | NP_780454.1 | 2 | 244 | ||
GenBank mRNA | AK010788.1 | - | 2 | 220 | ||
AK082817.1 | - | 2 | 190 | |||
AK087564.1 | - | 2 | 188 | |||
AK150268.1 | - | 2 | 220 | |||
AK151232.1 | - | 2 | 220 | |||
AK151358.1 | - | 2 | 220 | |||
AK151819.1 | - | 2 | 220 | |||
AK160911.1 | - | 2 | 229 | |||
AK165605.1 | - | 2 | 207 | |||
AK168114.1 | - | 2 | 185 | |||
AK169036.1 | - | 2 | 194 | |||
AK170746.1 | - | 2 | 194 | |||
AK209030.1 | - | 1 | 70 | |||
BB844588.1 | - | 2 | 245 | |||
BC038168.1 | - | 2 | 182 | |||
BC117005.1 | - | 2 | 219 | 40126324 | ||
BC117007.1 | - | 2 | 219 | 40126326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
