siRNA Details
Gene Symbol: 2410131K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081236.1 | NP_001074705.1 | 1 | 192 | ||
GenBank mRNA | AK010791.1 | - | 1 | 169 | ||
AK090380.1 | - | 1 | 151 | |||
BC104372.1 | - | 1 | 126 | 40049898 | ||
BC104373.1 | - | 1 | 126 | 40049899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
