siRNA Details
Gene Symbol: NF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000267.3 | NP_000258.1 | 12 | 1698 | ||
NM_001042492.2 | NP_001035957.1 | 12 | 1698 | |||
NM_001128147.2 | NP_001121619.1 | 12 | 1698 | |||
XM_005257983.2 | XP_005258040.1 | 12 | 1648 | |||
XM_005257984.2 | XP_005258041.1 | 12 | 1648 | |||
XM_006721922.2 | XP_006721985.1 | 13 | 1678 | |||
XM_006721924.2 | XP_006721987.1 | 13 | 1678 | |||
XM_006721925.2 | XP_006721988.1 | 13 | 1678 | |||
XM_006721926.3 | XP_006721989.1 | 13 | 1678 | |||
XM_011524852.2 | XP_011523154.1 | 13 | 1678 | |||
XM_011524857.2 | XP_011523159.1 | 13 | 1678 | |||
XM_017024689.1 | XP_016880178.1 | 13 | 1678 | |||
XM_017024690.1 | XP_016880179.1 | 13 | 1678 | |||
GenBank mRNA | AK289936.1 | - | 12 | 1648 | ||
BC144643.1 | - | 12 | 1417 | |||
BX537850.1 | - | 10 | 1090 | |||
D12625.1 | - | 12 | 1698 | |||
D42072.1 | - | 12 | 1380 | |||
M38106.1 | - | 4 | 313 | |||
M38107.1 | - | 4 | 313 | |||
M82814.1 | - | 12 | 1526 | |||
M89914.1 | - | 12 | 1512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
