siRNA Details
Gene Symbol: Rpl18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029751.4 | NP_084027.1 | 2 | 146 | ||
GenBank mRNA | AK010983.1 | - | 2 | 117 | ||
AK146543.1 | - | 2 | 116 | |||
AK150820.1 | - | 2 | 115 | |||
AK152611.1 | - | 2 | 115 | |||
AK161942.1 | - | 2 | 116 | |||
AK162047.1 | - | 2 | 117 | |||
BC024609.1 | - | 2 | 103 | 4221597 | ||
BC037146.1 | - | 2 | 158 | 5011713 | ||
CJ042325.1 | - | 2 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
