siRNA Details
Gene Symbol: NF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000267.3 | NP_000258.1 | 29 | 4292 | ||
NM_001042492.2 | NP_001035957.1 | 29 | 4292 | |||
XM_005257983.2 | XP_005258040.1 | 29 | 4242 | |||
XM_005257984.2 | XP_005258041.1 | 29 | 4242 | |||
XM_006721922.2 | XP_006721985.1 | 30 | 4272 | |||
XM_006721924.2 | XP_006721987.1 | 30 | 4272 | |||
XM_006721925.2 | XP_006721988.1 | 30 | 4272 | |||
XM_006721926.3 | XP_006721989.1 | 30 | 4272 | |||
XM_011524852.2 | XP_011523154.1 | 30 | 4269 | |||
XM_011524857.2 | XP_011523159.1 | 30 | 4272 | |||
XM_017024689.1 | XP_016880178.1 | 30 | 4272 | |||
XM_017024690.1 | XP_016880179.1 | 30 | 4272 | |||
GenBank mRNA | AB209336.1 | - | 12 | 2445 | ||
BC144643.1 | - | 29 | 4011 | |||
BX537850.1 | - | 27 | 3684 | |||
M38106.1 | - | 21 | 2907 | |||
M38107.1 | - | 21 | 2907 | |||
M60915.1 | - | 3 | 408 | |||
M61213.1 | - | 5 | 642 | |||
M82814.1 | - | 29 | 4120 | |||
M89914.1 | - | 29 | 4106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
