siRNA Details
Gene Symbol: Calcoco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271018.1 | NP_001257947.1 | 4, 5 | 430 | ||
GenBank mRNA | AF490344.1 | - | 4, 5 | 428 | ||
AK010816.1 | - | 4, 5 | 428 | |||
AK145542.1 | - | 4, 5 | 431 | |||
AK166710.1 | - | 4, 5 | 431 | |||
AK166756.1 | - | 4, 5 | 431 | |||
AK166939.1 | - | 4, 5 | 428 | |||
AK167009.1 | - | 4, 5 | 431 | |||
AK167172.1 | - | 4, 5 | 431 | |||
AK167291.1 | - | 4, 5 | 430 |
