siRNA Details
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029760.2 | NP_084036.2 | 2 | 170 | ||
GenBank mRNA | AK010827.1 | - | 2 | 152 | ||
AK083643.1 | - | 2 | 160 | |||
AK141257.1 | - | 2 | 134 | |||
AK152377.1 | - | 2 | 171 | |||
BC062175.1 | - | 2 | 151 | 30291107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
