siRNA Details
Gene Symbol: Gper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029771.3 | NP_084047.2 | 3 | 1067 | ||
GenBank mRNA | AK018203.1 | - | 3 | 971 | ||
AK030375.1 | - | 3 | 1068 | |||
BC138598.1 | - | 3 | 922 | 8861618 | ||
BC138616.1 | - | 3 | 922 | 8861636 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
