siRNA Details
Gene Symbol: Rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122685.1 | NP_001116157.1 | 6 | 1081 | ||
NM_029777.3 | NP_084053.3 | 6 | 903 | |||
GenBank mRNA | AK029751.1 | - | 6 | 886 | ||
AK076751.1 | - | 6 | 1082 | |||
AK079448.1 | - | 6 | 866 | |||
AK142277.1 | - | 6 | 892 | |||
AK161153.1 | - | 6 | 859 | |||
AY973322.1 | - | 5 | 754 | |||
BC005518.1 | - | 5 | 843 | 3494511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
