siRNA Details
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012340.4 | NP_036472.2 | 1, 2 | 350 | ||
NM_173091.3 | NP_775114.1 | 1, 2 | 350 | |||
XM_011528824.2 | XP_011527126.1 | 1, 2 | 341 | |||
XM_017027851.1 | XP_016883340.1 | 1, 2 | 341 | |||
GenBank mRNA | BC136418.1 | - | 1, 2 | 253 | 9020406 | |
BC144074.1 | - | 1, 2 | 253 | 9052594 | ||
CR993438.1 | - | 1, 2 | 325 | |||
EU887573.1 | - | 1, 2 | 130 | |||
EU887578.1 | - | 1, 2 | 130 | |||
U43341.1 | - | 1, 2 | 350 | |||
U43342.1 | - | 1, 2 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
