siRNA Details
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136021.2 | NP_001129493.1 | 9 | 2790 | ||
NM_001258292.1 | NP_001245221.1 | 9 | 2790 | |||
NM_001258294.1 | NP_001245223.1 | 9 | 2276 | |||
NM_001258295.1 | NP_001245224.1 | 9 | 2276 | |||
NM_001258296.1 | NP_001245225.1 | 9 | 2354 | |||
NM_001258297.1 | NP_001245226.1 | 9 | 2354 | |||
NM_012340.4 | NP_036472.2 | 9 | 2868 | |||
NM_173091.3 | NP_775114.1 | 9 | 2868 | |||
XM_011528824.2 | XP_011527126.1 | 9 | 2859 | |||
XM_011528825.2 | XP_011527127.1 | 9 | 2816 | |||
XM_011528826.2 | XP_011527128.1 | 9 | 2297 | |||
XM_017027850.1 | XP_016883339.1 | 9 | 2092 | |||
GenBank mRNA | AK226146.1 | - | 9 | 2790 | ||
BC136418.1 | - | 9 | 2771 | 9020406 | ||
BC144074.1 | - | 9 | 2771 | 9052594 | ||
BF914609.1 | - | 1 | 310 | |||
BU429325.1 | - | 1 | 194 | |||
EU887573.1 | - | 9 | 2648 | |||
EU887574.1 | - | 9 | 2588 | |||
EU887575.1 | - | 9 | 2276 | |||
EU887576.1 | - | 9 | 2354 | |||
EU887577.1 | - | 9 | 2588 | |||
EU887578.1 | - | 9 | 2648 | |||
EU887579.1 | - | 9 | 2276 | |||
EU887580.1 | - | 9 | 2354 | |||
U43341.1 | - | 9 | 2868 | |||
U43342.1 | - | 9 | 2868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
