siRNA Details
Gene Symbol: Arrdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029799.1 | NP_084075.1 | 1 | 225 | ||
GenBank mRNA | BC100496.1 | - | 1 | 225 | 30916615 |

More Information
Other Species Targeted

Gene Ontology Categories:
