siRNA Details
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101.3 | NP_001092.1 | 6 | 1762 | ||
GenBank mRNA | AK025375.1 | - | 6 | 1761 | ||
AK058019.1 | - | 6 | 1762 | |||
AK098751.1 | - | 7 | 1565 | |||
AK130062.1 | - | 6 | 1764 | |||
AK130157.1 | - | 6 | 1763 | |||
AK222925.1 | - | 6 | 1765 | |||
AK223032.1 | - | 6 | 1760 | |||
AK225414.1 | - | 6 | 1597 | |||
AK309997.1 | - | 7 | 1804 | |||
BC001301.1 | - | 6 | 1745 | 3451917 | ||
BC002409.2 | - | 6 | 1750 | 2961617 | ||
BC004251.1 | - | 6 | 1739 | 3623067 | ||
BC008633.1 | - | 5 | 1659 | |||
BC009636.1 | - | 1 | 496 | |||
BC012854.1 | - | 5 | 1635 | |||
BC013380.2 | - | 6 | 1754 | 4025526 | ||
BC013835.1 | - | 6 | 1751 | |||
BC014861.1 | - | 6 | 1752 | 3870487 | ||
BC016045.1 | - | 6 | 1753 | 4564532 | ||
X00351.1 | - | 6 | 1718 | |||
X63432.1 | - | 6 | 1750 |
Gene Symbol: SNORD140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132979.1 | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
