siRNA Details
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029804.3 | NP_084080.1 | 6 | 629 | ||
GenBank mRNA | AK011521.1 | - | 6 | 575 | ||
AK048220.1 | - | 1 | 1210 | |||
AK133911.1 | - | 6 | 564 | |||
AK166670.1 | - | 6 | 564 | |||
BC054785.1 | - | 2031 |

More Information
Other Species Targeted

Gene Ontology Categories:
