siRNA Details
Gene Symbol: Rbm17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152824.1 | NP_690037.1 | 9 | 1033 | ||
GenBank mRNA | AF083384.1 | - | 8 | 903 | ||
AK012296.1 | - | 9 | 979 | |||
BC034896.1 | - | 9 | 1033 | 1446602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
